Learn More
Enterprise Technology Solutions Betco Corporation Cleanser, Cream, Jeweler-grade Abrasive, 32oz, White
SDP
Catalog No. 5000790098
Encompass_Preferred
Best Bet Liquid Abrasive Creme Cleanser quickly removes embedded rust, soap scum, stains, grease, oxidation, hard water minerals and filth from a variety of surfaces. This liquid cream cleanser contains jeweler-grade abrasive that won't scratch surfaces like scouring powders. White, stable emulsion contains bleaching compounds, detergents and wetting agents. Thick, clinging formula is perfect for vertical surfaces. Use to clean bathtubs, chrome, stainless steel, porcelain, sinks, tile, Fiberglas enamel and a variety of other water-washable surfaces. It penetrates, brightens and polishes without heavy agitation. Cleanser is ready to use and pleasantly scented with a fresh mint fragrance.
Catalog No. 50-007-90098 Supplier Enterprise Technology Solutions Supplier No. BET0771200
Best Bet Liquid Abrasive Creme Cleanser quickly removes embedded rust, soap scum, stains, grease, oxidation, hard water minerals and filth from a variety of surfaces. This liquid cream cleanser contains jeweler-grade abrasive that won't scratch surfaces like scouring powders. White, stable emulsion contains bleaching compounds, detergents and wetting agents. Thick, clinging formula is perfect for vertical surfaces. Use to clean bathtubs, chrome, stainless steel, porcelain, sinks, tile, Fiberglas enamel and a variety of other water-washable surfaces. It penetrates, brightens and polishes without heavy agitation. Cleanser is ready to use and pleasantly scented with a fresh mint fragrance.