Learn More
Enterprise Technology Solutions Betco Corporation Degreaser, Bio-based, Concentrated, 1 Gallon, Dark Green
SDP
Catalog No. 5000790607
Encompass_Preferred
This powerful degreaser is the naturally preferred solution for all degreasing tasks. A synergistic blend of natural ingredients with a unique surfactant system produces a concentrated formula that equals most traditional degreasers. This unique degreaser breaks down greases, oils and fats including industrial petroleum greases on all water-safe surfaces. Green Earth Natural Degreaser removes inks, oils, greases and fats from a variety of surfaces with a proprietary blend of natural, bio-renewable/bio-based ingredients. Dilution ratio is 1:64.
Catalog No. 50-007-90607 Supplier Enterprise Technology Solutions Supplier No. BET2170400
This powerful degreaser is the naturally preferred solution for all degreasing tasks. A synergistic blend of natural ingredients with a unique surfactant system produces a concentrated formula that equals most traditional degreasers. This unique degreaser breaks down greases, oils and fats including industrial petroleum greases on all water-safe surfaces. Green Earth Natural Degreaser removes inks, oils, greases and fats from a variety of surfaces with a proprietary blend of natural, bio-renewable/bio-based ingredients. Dilution ratio is 1:64.