Learn More
Enterprise Technology Solutions Betco Corporation Degreaser, Bio-based, Concentrated, FastDraw, 2 Liter, DGN
SDP
Catalog No. 5000790137
Encompass_Preferred
This powerful degreaser is the naturally preferred solution for all degreasing tasks. A synergistic blend of natural ingredients with a unique surfactant system produces a concentrated formula that equals most traditional degreasers. This unique degreaser breaks down greases, oils and fats including industrial petroleum greases on all water-safe surfaces. Green Earth Natural Degreaser removes inks, oils, greases and fats from a variety of surfaces with a proprietary blend of natural, bio-renewable/bio-based ingredients. Dilution ratio is 1:64. As part of the FastDraw system, it is numbered (FastDraw 13) and color-coded to eliminate misuse. Spill-proof bottle helps prevent chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.
Catalog No. 50-007-90137 Supplier Enterprise Technology Solutions Supplier No. BET2174700
Add to Cart
Add to Cart
This powerful degreaser is the naturally preferred solution for all degreasing tasks. A synergistic blend of natural ingredients with a unique surfactant system produces a concentrated formula that equals most traditional degreasers. This unique degreaser breaks down greases, oils and fats including industrial petroleum greases on all water-safe surfaces. Green Earth Natural Degreaser removes inks, oils, greases and fats from a variety of surfaces with a proprietary blend of natural, bio-renewable/bio-based ingredients. Dilution ratio is 1:64. As part of the FastDraw system, it is numbered (FastDraw 13) and color-coded to eliminate misuse. Spill-proof bottle helps prevent chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.