Learn More
Enterprise Technology Solutions Betco Corporation Detergent,Dye/Fragrance-free,Concentrated,5 Gal,Clear
SDP
Catalog No. 5000790101
Encompass_Preferred
Count on the same great cleaning performance as Betco's Symplicity Detergent but without dyes and fragrance that cause allergies and sensitivities in some customers. Concentrated synthetic detergent with powerful surfactants and anti-redeposition agents release heavy soils from the linen, preventing dulling and leaving linen fresh and white. Blended surfactant technology means it is effective on a variety of soil types. Use as a standalone detergent or with Symplicity Break Alkaline Booster for the worst soils and stains. Detergency is Cold Water Effective, saving energy by getting the job done in cold water. For low environmental impact, it contains no phosphorous, APE, NPE or EDTA.
Catalog No. 50-007-90101 Supplier Enterprise Technology Solutions Supplier No. BET4987800
Count on the same great cleaning performance as Betco's Symplicity Detergent but without dyes and fragrance that cause allergies and sensitivities in some customers. Concentrated synthetic detergent with powerful surfactants and anti-redeposition agents release heavy soils from the linen, preventing dulling and leaving linen fresh and white. Blended surfactant technology means it is effective on a variety of soil types. Use as a standalone detergent or with Symplicity Break Alkaline Booster for the worst soils and stains. Detergency is Cold Water Effective, saving energy by getting the job done in cold water. For low environmental impact, it contains no phosphorous, APE, NPE or EDTA.