Learn More
Enterprise Technology Solutions Betco Corporation Detergent, f/Pots&Pans, Heavy-duty, FastDraw, 2 Liter, YW
SDP
Catalog No. 5000790606
Encompass_Preferred
Heavy-duty detergent is specifically designed for restaurant, institutional and industrial use where long-lasting suds and easy removal of grease are a must. It is ideal for cleaning pots, pans and dishes. This lemon-scented detergent works quickly and leaves surfaces spot-free and film-free. Symplicity Citrusuds is made from the best surfactants and rinses quickly and easily. It is effective on grease and fat yet mild on the hands. As part of the FastDraw system, it is numbered (FastDraw 26) and color-coded to eliminate misuse. NAFTA trilingual labels are suited to a multicultural workforce. Spill-proof bottle design eliminates the potential for chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Metering insert reduces end-use cost per gallon by delivering accurate dilutions. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.
Catalog No. 50-007-90606 Supplier Enterprise Technology Solutions Supplier No. BET21094700
Heavy-duty detergent is specifically designed for restaurant, institutional and industrial use where long-lasting suds and easy removal of grease are a must. It is ideal for cleaning pots, pans and dishes. This lemon-scented detergent works quickly and leaves surfaces spot-free and film-free. Symplicity Citrusuds is made from the best surfactants and rinses quickly and easily. It is effective on grease and fat yet mild on the hands. As part of the FastDraw system, it is numbered (FastDraw 26) and color-coded to eliminate misuse. NAFTA trilingual labels are suited to a multicultural workforce. Spill-proof bottle design eliminates the potential for chemical contact with cleaning staff. Patented technology uses two metering tips for low-flow and high-flow dilution. Metering insert reduces end-use cost per gallon by delivering accurate dilutions. Curved siphon tubes on the FastDraw Dispensers (not included) allow complete product evacuation, eliminating waste.