Learn More
Enterprise Technology Solutions Betco Corporation Detergent,Machine,Conc Liquid,All-Temp,5 Gal Pail,CL OE
SDP
Catalog No. 5000790276
Encompass_Preferred
Symplicity All Temp Machine Detergent delivers superior dish-cleaning power and water hardness management on a wide range of soils. Clear orange, liquid formulation is optimized for water-conditioning soil suspension and rinsability. Heavy-duty, nonchlorinated detergent assures instant mixing consistent results and automatic dispensing ease. Use in both high-temp or low-temp machines. Concentrated, machine-warewashing detergent is also hard water-effective. It is NSF International Certified for use in all regulated food-processing and foodservice environments. Sure Connect dispensing eliminates waste and employee guesswork with a closed-loop system that prevents accidental spills.
Catalog No. 50-007-90276 Supplier Enterprise Technology Solutions Supplier No. BET2447800
Symplicity All Temp Machine Detergent delivers superior dish-cleaning power and water hardness management on a wide range of soils. Clear orange, liquid formulation is optimized for water-conditioning soil suspension and rinsability. Heavy-duty, nonchlorinated detergent assures instant mixing consistent results and automatic dispensing ease. Use in both high-temp or low-temp machines. Concentrated, machine-warewashing detergent is also hard water-effective. It is NSF International Certified for use in all regulated food-processing and foodservice environments. Sure Connect dispensing eliminates waste and employee guesswork with a closed-loop system that prevents accidental spills.