Learn More
Enterprise Technology Solutions Betco Corporation Dishwashing Detergent, Manual, Liquid, 1Gal, Blue
SDP
Catalog No. 5000790113
Encompass_Preferred
Ultra-concentrated, dishwashing detergent quickly leaves dishes sparkling clean and film-free during manual dishwashing. Ideal for cleaning heavily encrusted pots and pans, it has the power to cut through the toughest grease. Simplicity In-Sync Hand Dishwashing Detergent is specifically designed for supermarket, institutional and restaurant use where fast grease emulsification and lasting suds are a must. Mountains of hardworking suds penetrate grease and food deposits, leaving pots, pans, glassware and china spot-free. Premium detergent rinses freely and never streaks. It is mild and gentle to hands at dilution. Concentrated formula only uses 1 to 2 oz. per 10 gallons of water.
Catalog No. 50-007-90113 Supplier Enterprise Technology Solutions Supplier No. BET18510400
Ultra-concentrated, dishwashing detergent quickly leaves dishes sparkling clean and film-free during manual dishwashing. Ideal for cleaning heavily encrusted pots and pans, it has the power to cut through the toughest grease. Simplicity In-Sync Hand Dishwashing Detergent is specifically designed for supermarket, institutional and restaurant use where fast grease emulsification and lasting suds are a must. Mountains of hardworking suds penetrate grease and food deposits, leaving pots, pans, glassware and china spot-free. Premium detergent rinses freely and never streaks. It is mild and gentle to hands at dilution. Concentrated formula only uses 1 to 2 oz. per 10 gallons of water.