Learn More
Enterprise Technology Solutions Betco Corporation Disinfectant, Hospital-Grade, No Rinse, 32 oz, Clear
SDP
Catalog No. 5000832716
Encompass_Preferred
Disinfectant with the environment in mind using GE Fight Bac RTU. It kills more than 99.9 percent of bacteria and viruses with a 3-to-5-minute contact time. This hospital-grade product provides superior disinfection and meets the EPA's Design for the Environment standards. GE Fight Bac RTU is an EPA list N disinfectant. All products on this list meet EPA's criteria for use against SARS-CoV-2, the virus that causes COVID-19. GE Fight Bac RTU is formulated with a botanical active ingredient: citric acid. Citric acid is nonirritating to most and emits no harsh fumes for the user. This disinfectant is also free from harsh chemicals so it can be used as a food contact sanitizer with no rinsing or wiping required. Use it on washable hard nonporous in hospitals, kitchens, bathrooms, healthcare facilities, medical offices, daycare centers, schools, nursing homes and grocery stores.
Catalog No. 50-008-32716 Supplier Enterprise Technology Solutions Supplier No. BET3901200
Disinfectant with the environment in mind using GE Fight Bac RTU. It kills more than 99.9 percent of bacteria and viruses with a 3-to-5-minute contact time. This hospital-grade product provides superior disinfection and meets the EPA's Design for the Environment standards. GE Fight Bac RTU is an EPA list N disinfectant. All products on this list meet EPA's criteria for use against SARS-CoV-2, the virus that causes COVID-19. GE Fight Bac RTU is formulated with a botanical active ingredient: citric acid. Citric acid is nonirritating to most and emits no harsh fumes for the user. This disinfectant is also free from harsh chemicals so it can be used as a food contact sanitizer with no rinsing or wiping required. Use it on washable hard nonporous in hospitals, kitchens, bathrooms, healthcare facilities, medical offices, daycare centers, schools, nursing homes and grocery stores.