Learn More
Enterprise Technology Solutions Betco Corporation Dispenser f/4 Cleaning Fluids, ActionGap, Bottle/Bucket
SDP
Supplier:  Enterprise Technology Solutions BET9217400
Encompass_Preferred
This four-bay, locking, chemical management system handles the toughest environments for convenient dispensing of ready-to-use cleaning fluids. Durable looking exterior is matched by a robust three-point lock for each door to prevent tampering or theft and hinges that withstand the toughest cleaning environment. FastDraw Pro Dispenser (ActionGap) is designed to dispense cleaning solutions mixed with water to bottles or buckets from each bay and can be mounted in a corner or flat walls. It uses an ActionGap mechanism to prevent backflow and contamination of water supply. Filling quart bottles has never been simpler. Slide the fill tube into the bottle and push up. Four bays mean you can stock it with four different cleaning solutions mostly frequently used by your team. Space-saving design mounts on either a flat wall or in a corner.
Catalog No. 50-008-10489