Learn More
Enterprise Technology Solutions Betco Corporation Dispenser,f/Chemicals,w/Lock,2 FlowRates,13"x25.75"x7.25",GY
SDP
Catalog No. 5000810508
Encompass_Preferred
FastDraw PRO Action Gap delivers easier, more reliable and convenient chemical management. Smaller, lighter design requires less wall space and fits in tight spaces. Hidden drip tray catches accidental product drips. Eight-inch discharge hose makes filling automatic scrubbers easier. Durable exterior and robust three-point door lock provide extra protection for sensitive chemicals. All internal parts can be accessed by convenient one-door design. Easily fill bottles and buckets from all bays. Two independent flow rates (one GPM and four GPM) ensure you always ensure correct dilution.
Catalog No. 50-008-10508 Supplier Enterprise Technology Solutions Supplier No. BET9268200
FastDraw PRO Action Gap delivers easier, more reliable and convenient chemical management. Smaller, lighter design requires less wall space and fits in tight spaces. Hidden drip tray catches accidental product drips. Eight-inch discharge hose makes filling automatic scrubbers easier. Durable exterior and robust three-point door lock provide extra protection for sensitive chemicals. All internal parts can be accessed by convenient one-door design. Easily fill bottles and buckets from all bays. Two independent flow rates (one GPM and four GPM) ensure you always ensure correct dilution.