Learn More
Enterprise Technology Solutions Betco Corporation Laundry Detergent, Liquid, Concentrated, 1 Gal, 4/CT, Blue
SDP
Catalog No. 5000790138
Encompass_Preferred
Symplicity Duet L Liquid Laundry Detergent brightens and whitens as it safely lifts stubborn dirt and stains. This all-in-one detergent with enzymes performs well in all water temperatures and is safe for all types of water-fast natural and synthetic fabrics. It is even with a chlorine scavenger to battle fading fabric by preventing chlorine in the water supply. Concentrated formula saves time and delivers cost-effective use. It makes low environmental impact with low VOCs and no phosphorous, APE, NPE or EDTA.
Catalog No. 50-007-90138 Supplier Enterprise Technology Solutions Supplier No. BET4750400
Symplicity Duet L Liquid Laundry Detergent brightens and whitens as it safely lifts stubborn dirt and stains. This all-in-one detergent with enzymes performs well in all water temperatures and is safe for all types of water-fast natural and synthetic fabrics. It is even with a chlorine scavenger to battle fading fabric by preventing chlorine in the water supply. Concentrated formula saves time and delivers cost-effective use. It makes low environmental impact with low VOCs and no phosphorous, APE, NPE or EDTA.