Learn More
Enterprise Technology Solutions Betco Corporation Restroom Cleaner,Conc,Hvy-dty,FastDraw,1/2 Gal (2L),4/CT
SDP
Catalog No. 5000832222
Encompass_Preferred
Heavy-duty cleaner is chemically designed to remove heavy soap scum, body and tanning oils, mineral deposits and other soils found in all areas of the restroom. It is effective on tile and grout surfaces, countertops, sinks and toilets. High-foaming action allows longer contact time and maximum penetration of tough soils. Use Green Earth Restroom Cleaner on acid-resistant surfaces such as showers, bathtubs and other restroom surfaces. Citrus Floral scent leaves a pleasant, powerful fragrance. Do not use on marble, aluminum, terrazzo, Formica or carpeting. As part of the FastDraw system, the highly concentrated sanitizer in a spill-proof bottle is numbered (FastDraw 32) and color-coded to eliminate misuse. Metering insert reduces end-use cost per gallon by delivering accurate dilutions.
Catalog No. 50-008-32222 Supplier Enterprise Technology Solutions Supplier No. BET5484700
Heavy-duty cleaner is chemically designed to remove heavy soap scum, body and tanning oils, mineral deposits and other soils found in all areas of the restroom. It is effective on tile and grout surfaces, countertops, sinks and toilets. High-foaming action allows longer contact time and maximum penetration of tough soils. Use Green Earth Restroom Cleaner on acid-resistant surfaces such as showers, bathtubs and other restroom surfaces. Citrus Floral scent leaves a pleasant, powerful fragrance. Do not use on marble, aluminum, terrazzo, Formica or carpeting. As part of the FastDraw system, the highly concentrated sanitizer in a spill-proof bottle is numbered (FastDraw 32) and color-coded to eliminate misuse. Metering insert reduces end-use cost per gallon by delivering accurate dilutions.