Learn More
Enterprise Technology Solutions Betco Corporation Rinse Aid,f/Dishwashers,Low&High Temp,Concentrated,4Gal,Blue
SDP
Catalog No. 5000832481
Encompass_Preferred
Symplicity Machine Rinse Aid delivers rapid sheeting for spot-free drying in both low and high temperature dishwashers. Low-foaming surfactant formula provides superior drying performance and eliminates alkaline and hard-water deposits.
Catalog No. 50-008-32481 Supplier Enterprise Technology Solutions Supplier No. BET2617400
Symplicity Machine Rinse Aid delivers rapid sheeting for spot-free drying in both low and high temperature dishwashers. Low-foaming surfactant formula provides superior drying performance and eliminates alkaline and hard-water deposits.