Learn More
Enterprise Technology Solutions Betco Corporation Rinse Aid,f/Dishwashing Machines,Conc,5 Gal Pail, Blue
SDP
Supplier:  Enterprise Technology Solutions BET2617800
Encompass_Preferred
Symplicity Machine Rinse Aid delivers rapid sheeting for spot-free drying in both low-temp and high-temp dish machines. Low-foaming surfactant formula provides superior drying performance. Its excellent sheeting action assists in eliminating alkaline and hard water deposits. Rinse additive is highly concentrated. Sure Connect dispensing eliminates waste and employee guesswork with a closed-loop system that prevents accidental spills.
Catalog No. 50-008-32833