Learn More
Enterprise Technology Solutions Betco Corporation Sanitizer, Concentrated, FastDraw 18, 2 Liter, Pink
SDP
Catalog No. 5000790135
Encompass_Preferred
EPA-approved, broad-range sanitizer delivers correct sanitizing at a range of 150 to 400 ppm. That allows the sanitizing solution to adapt to varying water-pressure situations while maintaining compliance with health department regulations. Multi-Range Sanibet kills 99.99 percent of foodservice organisms (listeria, E. coli and salmonella). It can be used to sanitize serving ware, utensils, pots and pans, and food-processing equipment. It can also be used for disinfecting floors and walls. This complete, chemically balanced deodorizer/disinfectant/sanitizer is effective even in the presence of hard water. As part of the FastDraw system, the highly concentrated sanitizer in a spill-proof bottle is numbered (FastDraw 18) and color-coded to eliminate misuse. Metering insert reduces end-use cost per gallon by delivering accurate dilutions.
Catalog No. 50-007-90135 Supplier Enterprise Technology Solutions Supplier No. BET2374700
EPA-approved, broad-range sanitizer delivers correct sanitizing at a range of 150 to 400 ppm. That allows the sanitizing solution to adapt to varying water-pressure situations while maintaining compliance with health department regulations. Multi-Range Sanibet kills 99.99 percent of foodservice organisms (listeria, E. coli and salmonella). It can be used to sanitize serving ware, utensils, pots and pans, and food-processing equipment. It can also be used for disinfecting floors and walls. This complete, chemically balanced deodorizer/disinfectant/sanitizer is effective even in the presence of hard water. As part of the FastDraw system, the highly concentrated sanitizer in a spill-proof bottle is numbered (FastDraw 18) and color-coded to eliminate misuse. Metering insert reduces end-use cost per gallon by delivering accurate dilutions.