Learn More
Enterprise Technology Solutions Betco Corporation Skin Cleanser, Foaming, Antibacterial, 500 ml, Light Blue
SDP
Catalog No. 5000810571
Encompass_Preferred
Ultra-Blue Antibacterial Foaming Skin Cleanser contains 0.13 percent benzalkonium chloride for rapid broad-spectrum germ-kill. It meets FDA and CDC requirements for handwashing. The quick-lathering cleanser is gentle on skin, moisturizing while leaving a pleasant fragrance (Clean Ocean). It will not cause dryness or skin irritation, encouraging frequent handwashing. Skin cleanser comes in a pump bottle for easy dispensing.
Catalog No. 50-008-10571 Supplier Enterprise Technology Solutions Supplier No. BET7590900
Ultra-Blue Antibacterial Foaming Skin Cleanser contains 0.13 percent benzalkonium chloride for rapid broad-spectrum germ-kill. It meets FDA and CDC requirements for handwashing. The quick-lathering cleanser is gentle on skin, moisturizing while leaving a pleasant fragrance (Clean Ocean). It will not cause dryness or skin irritation, encouraging frequent handwashing. Skin cleanser comes in a pump bottle for easy dispensing.