Learn More
Enterprise Technology Solutions Betco Corporation Soap Dispenser,Foam,Touch-Free,WE
SDP
Supplier:  Enterprise Technology Solutions BET9302600
Encompass_Preferred
Clario C3 Foam Touch-Free Dispensing System combines a sleek, modern design with advanced functionality in a hygienic closed-loop system. Its permanent pump reduces waste by 94 percent and lasts through 125 refills, delivering thick, rich foam with three adjustable shot sizes of 0.5 ml., 1.0 ml. and 1.5 ml. for precise dispensing. Sealed refills (not included) prevent cross-contamination, and 98 percent evacuation ensures more usable product per refill. It features an industry-leading battery life of 150,000 shots or four years (batteries included) and offers options for locked or open refill access. Fully ADA compliant, the Clario C3 is a practical, efficient, and safe touch-free solution for hand hygiene needs, continuing the legacy of value associated with the Clario name.
Catalog No. 50-008-32607