Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BETd-246 100mg | 2140289-17-2 | 946.02 g/mol | C48H55N11O10 | 100 MG
SDP

Catalog No. 5000226637
Encompass_Preferred

BETd-246 is a second-generation PROTAC-based BET bromodomain degrader that links Cereblon and BET ligands to induce selective degradation of BRD2, BRD3, and BRD4. It is intended for research use in biochemical, cell-based, and preclinical studies and has supporting solubility, purity, and activity data suitable for experimental applications.

  • Second-generation PROTAC degrader targeting BET family proteins (BRD2/BRD3/BRD4).
  • High reported purity of 98.34%.
  • Molecular weight 946.02 g/mol.
  • Soluble in DMSO at 200 mg/mL; in vivo formulation example: 10% DMSO/90% corn oil (≥ 5 mg/mL).
  • Demonstrates potent cellular activity (e.g., MDA-MB-468 IC50 < 10 nM; Burkitt's lymphoma GI50 0.4 μM).
  • Recommended storage: -20°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in research-scale quantities including 100 mg.

Catalog No. 50-002-26637 Supplier Medchemexpress LLC Supplier No. HY115568100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BETd-246 is a second-generation PROTAC-based BET bromodomain degrader that links Cereblon and BET ligands to induce selective degradation of BRD2, BRD3, and BRD4. It is intended for research use in biochemical, cell-based, and preclinical studies and has supporting solubility, purity, and activity data suitable for experimental applications.

  • Second-generation PROTAC degrader targeting BET family proteins (BRD2/BRD3/BRD4).
  • High reported purity of 98.34%.
  • Molecular weight 946.02 g/mol.
  • Soluble in DMSO at 200 mg/mL; in vivo formulation example: 10% DMSO/90% corn oil (≥ 5 mg/mL).
  • Demonstrates potent cellular activity (e.g., MDA-MB-468 IC50 < 10 nM; Burkitt's lymphoma GI50 0.4 μM).
  • Recommended storage: -20°C under nitrogen; in solvent: -80°C (6 months) or -20°C (1 month).
  • Available in research-scale quantities including 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.