Learn More
Medchemexpress LLC HY-115568 10mg Medchemexpress, BETd-246 CAS:2140289-17-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11556810MG
Encompass_Preferred
Medchemexpress, HY-115568 10mg BETd-246 CAS:2140289-17-2 BETd-246 is a second-generation and PROTAC-based BET bromodomain (BRD) inhibitor, exhibiting superior selectivity, potency and antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9393