Learn More
Medchemexpress LLC BETd-246 50mg | 2140289-17-2 | 946.02 | C48H55N11O10 | 50 MG
SDP
Catalog No. 5000228655
Encompass_Preferred
BETd-246 is a second-generation PROTAC-based BET bromodomain degrader that induces targeted degradation of BET family proteins (BRD2, BRD3, BRD4). It demonstrates high potency, selectivity, and antitumor activity in cellular assays and in vivo models. Supplied for research use only.
  • Enables targeted degradation of BET proteins.
  • High potency and selectivity in cellular assays.
  • Induces dose-dependent depletion of BRD2, BRD3, and BRD4.
  • Demonstrates antitumor efficacy in PDX models.
  • Soluble in DMSO at high concentration for in vitro use.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-28655 Supplier Medchemexpress LLC Supplier No. HY11556850MG
BETd-246 is a second-generation PROTAC-based BET bromodomain degrader that induces targeted degradation of BET family proteins (BRD2, BRD3, BRD4). It demonstrates high potency, selectivity, and antitumor activity in cellular assays and in vivo models. Supplied for research use only.
  • Enables targeted degradation of BET proteins.
  • High potency and selectivity in cellular assays.
  • Induces dose-dependent depletion of BRD2, BRD3, and BRD4.
  • Demonstrates antitumor efficacy in PDX models.
  • Soluble in DMSO at high concentration for in vitro use.
  • Stable when stored under recommended conditions.