Learn More
Medchemexpress LLC HY-115568 5mg Medchemexpress, BETd-246 CAS:2140289-17-2 Purity:>98%
SDP
Catalog No. 501932507
Encompass_Preferred
Medchemexpress, HY-115568 5mg BETd-246 CAS:2140289-17-2 BETd-246 is a second-generation BET bromodomain (BRD) inhibitor, exhibiting superior selectivity, potency and antitumor activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2507 Supplier Medchemexpress LLC Supplier No. HY1155685MG
Medchemexpress, HY-115568 5mg BETd-246 CAS:2140289-17-2 BETd-246 is a second-generation BET bromodomain (BRD) inhibitor, exhibiting superior selectivity, potency and antitumor activity [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.