Learn More
Selleck Chemical LLC Bethanechol chloride S2455-1g
SDP
Catalog No. 503111662
Encompass_Preferred
Bethanechol chloride(Myocholine Urecholine) is a selective muscarinic receptor agonist without any effect on nicotinic receptors
Catalog No. 50-311-1662 Supplier Selleck Chemical LLC Supplier No. S24551G
Bethanechol chloride(Myocholine Urecholine) is a selective muscarinic receptor agonist without any effect on nicotinic receptors