Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bethanechol (chloride) | 590-63-6 | 99.6% | 5 G
SDP

Catalog No. 5000409055
Encompass_Preferred

Bethanechol chloride acts as a parasympathomimetic agent and a mAChR agonist, directly stimulating all five muscarinic acetylcholine receptor subtypes (M1-M5) within the parasympathetic nervous system. Primarily for research use, this compound has demonstrated significant reduction in ileal pacemaker potentials in vitro. In vivo studies reveal that Bethanechol can induce drinking and increase urine output in rats in a dose-dependent manner. It has a molecular weight of 196.68 and a chemical formula of C7H17ClN2O2, presenting as a white to off-white solid. It exhibits solubility in water (≥ 50 mg/mL) and DMSO (11.11 mg/mL).

  • Directly stimulates mAChR (M1, M2, M3, M4, and M5)
  • Significantly reduces ileal pacemaker potentials in vitro
  • Induces drinking and increases urine output in rats in vivo

Catalog No. 50-004-09055 Supplier Medchemexpress LLC Supplier No. HYB0406A5G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bethanechol chloride acts as a parasympathomimetic agent and a mAChR agonist, directly stimulating all five muscarinic acetylcholine receptor subtypes (M1-M5) within the parasympathetic nervous system. Primarily for research use, this compound has demonstrated significant reduction in ileal pacemaker potentials in vitro. In vivo studies reveal that Bethanechol can induce drinking and increase urine output in rats in a dose-dependent manner. It has a molecular weight of 196.68 and a chemical formula of C7H17ClN2O2, presenting as a white to off-white solid. It exhibits solubility in water (≥ 50 mg/mL) and DMSO (11.11 mg/mL).

  • Directly stimulates mAChR (M1, M2, M3, M4, and M5)
  • Significantly reduces ileal pacemaker potentials in vitro
  • Induces drinking and increases urine output in rats in vivo

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.