Learn More
Medchemexpress LLC HY-103546 5mg Medchemexpress, BETP CAS:1371569-69-5 Purity:>98%
SDP
Catalog No. 501968127
Encompass_Preferred
Medchemexpress, HY-103546 5mg BETP CAS:1371569-69-5 BETP is an agonist of glucagon-like peptide-1 (GLP-1) receptor, with EC50s of 0.66 and 0.755 μM for human and rat GLP-1 receptor, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8127 Supplier Medchemexpress LLC Supplier No. HY1035465MG
Medchemexpress, HY-103546 5mg BETP CAS:1371569-69-5 BETP is an agonist of glucagon-like peptide-1 (GLP-1) receptor, with EC50s of 0.66 and 0.755 μM for human and rat GLP-1 receptor, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.