Learn More
Medchemexpress LLC Betrixaban | 330942-05-7 | MFCD16038040 | 451.91 g/mol | C23H22ClN5O3 | 5 MG
SDP
Catalog No. 501871992
Encompass_Preferred
Betrixaban is a selective, orally efficacious small-molecule inhibitor of factor Xa (fXa) supplied for research use. It exhibits potent inhibitory activity (IC50 = 1.5 nM) and is commonly used in biochemical assays and preclinical antithrombotic studies.
  • Potent fXa inhibition (IC50 = 1.5 nM).
  • Suitable for in vitro biochemical assays and preclinical antithrombotic models.
  • Research-grade compound supplied as a solid for analytical and experimental use.
  • CAS number 330942-05-7 and molecular formula C23H22ClN5O3.
  • Available in small pack sizes suitable for assay development (1 mg, 5 mg, 10 mg).
Catalog No. 50-187-1992 Supplier Medchemexpress LLC Supplier No. HY102685MG
Betrixaban is a selective, orally efficacious small-molecule inhibitor of factor Xa (fXa) supplied for research use. It exhibits potent inhibitory activity (IC50 = 1.5 nM) and is commonly used in biochemical assays and preclinical antithrombotic studies.
  • Potent fXa inhibition (IC50 = 1.5 nM).
  • Suitable for in vitro biochemical assays and preclinical antithrombotic models.
  • Research-grade compound supplied as a solid for analytical and experimental use.
  • CAS number 330942-05-7 and molecular formula C23H22ClN5O3.
  • Available in small pack sizes suitable for assay development (1 mg, 5 mg, 10 mg).