Learn More
Medchemexpress LLC Betrixaban-d6 | 2098655-51-5 | 99.0% | 457.94 g/mol | C23H16D6ClN5O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY10268S1MG
Encompass_Preferred
Betrixaban-d6 is a deuterium-labeled analog of betrixaban, provided for use as an internal standard and tracer in quantitative analytical workflows. The compound is intended for LC-MS, GC-MS, NMR, and other assays where isotopic labeling improves quantitation and peak identification.
  • Deuterium-labeled analog suitable for use as internal standard
  • High purity for reliable analytical results
  • Compatible with LC-MS, GC-MS, and NMR assays
  • Supplied in small quantities for analytical method development
Catalog No. 50-002-45760