Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Betrixaban maleate | 936539-80-9 | MFCD16038040 | 99.8% | 567.98 | C27H26ClN5O7 | 10 MG
SDP

Catalog No. 5000246931
Encompass_Preferred

Betrixaban maleate is the maleate salt of betrixaban, a potent and selective orally efficacious factor Xa (fXa) inhibitor with an IC50 of 1.5 nM. Supplied as a solid for research use, it demonstrates antithrombotic activity and includes specified purity, storage, and solubility data for laboratory handling.

  • High purity (99.8%).
  • White to off-white solid form.
  • Molecular weight 567.98 g/mol.
  • CAS number 936539-80-9.
  • Soluble in DMSO (50 mg/mL) and in water (2 mg/mL) with warming and sonication.
  • Storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month).
  • Provided as a 10 MG research pack.

Catalog No. 50-002-46931 Supplier Medchemexpress LLC Supplier No. HY10268A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Betrixaban maleate is the maleate salt of betrixaban, a potent and selective orally efficacious factor Xa (fXa) inhibitor with an IC50 of 1.5 nM. Supplied as a solid for research use, it demonstrates antithrombotic activity and includes specified purity, storage, and solubility data for laboratory handling.

  • High purity (99.8%).
  • White to off-white solid form.
  • Molecular weight 567.98 g/mol.
  • CAS number 936539-80-9.
  • Soluble in DMSO (50 mg/mL) and in water (2 mg/mL) with warming and sonication.
  • Storage: 4°C sealed; in solvent -80°C (6 months), -20°C (1 month).
  • Provided as a 10 MG research pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.