Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Betrixaban maleate | 936539-80-9 | MFCD31729381 | 99.8% | 567.98 g/mol | C27H26ClN5O7 | 25 MG
SDP

Catalog No. 5000245250
Encompass_Preferred

Betrixaban maleate is the maleate salt of betrixaban, a potent and selective orally active factor Xa (fXa) inhibitor used as a research compound to study coagulation and antithrombotic pharmacology. The compound is supplied as a high-purity research chemical suitable for in vitro and in vivo experiments.

  • High purity suitable for research (99.78%).
  • Maleate salt improves handling and solubility properties.
  • Targets factor Xa for antithrombotic studies.
  • Molecular weight 567.98 g/mol; formula C27H26ClN5O7.
  • Available in small milligram quantities for laboratory use (e.g., 25 mg).
  • Supplied for research use only; not for human or clinical applications.

Catalog No. 50-002-45250 Supplier Medchemexpress LLC Supplier No. HY10268A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Betrixaban maleate is the maleate salt of betrixaban, a potent and selective orally active factor Xa (fXa) inhibitor used as a research compound to study coagulation and antithrombotic pharmacology. The compound is supplied as a high-purity research chemical suitable for in vitro and in vivo experiments.

  • High purity suitable for research (99.78%).
  • Maleate salt improves handling and solubility properties.
  • Targets factor Xa for antithrombotic studies.
  • Molecular weight 567.98 g/mol; formula C27H26ClN5O7.
  • Available in small milligram quantities for laboratory use (e.g., 25 mg).
  • Supplied for research use only; not for human or clinical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.