Learn More
Selleck Chemical LLC Betrixaban maleate S5389-5mg
SDP
Catalog No. 503108561
Encompass_Preferred
Betrixaban maleate is the maleate salt form of Betrixaban which is a Factor Xa inhibitor that decreases prothrombinase activity and thrombin generation
Catalog No. 50-310-8561 Supplier Selleck Chemical LLC Supplier No. S53895MG
Betrixaban maleate is the maleate salt form of Betrixaban which is a Factor Xa inhibitor that decreases prothrombinase activity and thrombin generation