Learn More
AdipoGen Betulin, 473-98-3, MFCD00016802, 1 g
SDP
Catalog No. 501146391
Encompass_Preferred
MF: C30H50O2, MW: 442.7, ≥98%, Appearance: White solid. Specific SREBPs (sterol regulatory element-binding proteins) inhibitor. Inhibits ER-Golgi translocation of SREBPs through binding to SCAP in an INSIG-dependent manner.
Catalog No. 50-114-6391 Supplier AdipoGen Supplier No. AGCN20476G001
MF: C30H50O2, MW: 442.7, ≥98%, Appearance: White solid. Specific SREBPs (sterol regulatory element-binding proteins) inhibitor. Inhibits ER-Golgi translocation of SREBPs through binding to SCAP in an INSIG-dependent manner.