Learn More
Cayman Chemical Betulin 3,28-diacetate, Purity: ≥95%, CAS: 1721-69-3, 25 mg
SDP
Catalog No. 502948985
Encompass_Preferred
A triterpene with antiviral and hepatoprotective activities; inhibits SFV replication in virus-infected Huh7 cells (IC50 = 9.1 µM); improves hepatic function in various rat models of hepatitis at 20 mg/kg
Catalog No. 50-294-8985 Supplier Cayman Chemical Supplier No. 3302825 MG
A triterpene with antiviral and hepatoprotective activities; inhibits SFV replication in virus-infected Huh7 cells (IC50 = 9.1 µM); improves hepatic function in various rat models of hepatitis at 20 mg/kg