Learn More
AdipoGen Betulin
SDP
Supplier:  AdipoGen AGCN20476M250
Encompass_Preferred
Chemical. CAS 473-98-3. Formula C30H50O2. MW 442.7. Isolated from Betulaceae family tree bark. Specific SREBPs sterol regulatory element-binding proteins inhibitor. Inhibits ER-Golgi translocation of SREBPs through binding to SCAP in an INSIG-dependent manner. Improves hyperlipidemia and insulin resistance and reduces atherosclerotic plaques. Lowers cholesterol levels through inhibition of SREBP-2 activation. Reduces weight gain and adipose tissue size in vivo. Increases oxygen consumption and energy expenditure. Increases UCP1/2 levels in brown adipose tissue BAT after cold stimulation. Anticancer compound. Apoptosis inducer. DNA topoisomerase II inhibitor. Anti-inflammatory compound. Shown to inhibit LPS-induced NO production through the TLR4/NF-kappaB signaling pathway. Antiviral compound.
Catalog No. 50-256-2515