Learn More
Medchemexpress LLC HY-N0083 50mg Medchemexpress, Betulin CAS:473-98-3 Purity:>98%
SDP
Catalog No. 501933382
Encompass_Preferred
Medchemexpress, HY-N0083 50mg Betulin CAS:473-98-3 Betulin (Trochol), is a sterol regulatory element-binding protein ( SREBP ) inhibitor with an IC 50 of 14.5 μM in K562 cell line. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3382 Supplier Medchemexpress LLC Supplier No. HYN008350MG
Medchemexpress, HY-N0083 50mg Betulin CAS:473-98-3 Betulin (Trochol), is a sterol regulatory element-binding protein ( SREBP ) inhibitor with an IC 50 of 14.5 μM in K562 cell line. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.