Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betulin, 95.2%, MP Biomedicals™

Catalog No. ICN10100601 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10100601 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10100601 Supplier MP Biomedicals, Inc Supplier No. 0210100601
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 473-98-3
Molecular Formula C30H50O2
Molecular Weight (g/mol) 442.728
InChI Key FVWJYYTZTCVBKE-ROUWMTJPSA-N
Synonym betulin, betulinol, betuline, trochol, betulol, lup-20 29-ene-3b,28-diol, lup-20 29-ene-3beta,28-diol, unii-6w70hn7x7o, lup-20 30-ene-3beta,28-diol, lup-20 29-ene-3,28-diol, 3beta
PubChem CID 72326
ChEBI CHEBI:3086
IUPAC Name (1R,3aS,5aR,5bR,7aR,9S,11aR,11bR,13aR,13bR)-3a-(hydroxymethyl)-5a,5b,8,8,11a-pentamethyl-1-prop-1-en-2-yl-1,2,3,4,5,6,7,7a,9,10,11,11b,12,13,13a,13b-hexadecahydrocyclopenta[a]chrysen-9-ol
SMILES CC(=C)C1CCC2(C1C3CCC4C5(CCC(C(C5CCC4(C3(CC2)C)C)(C)C)O)C)CO

Specifications

Color White
Assay Percent Range 95.2%
Quantity 1 g
Formula Weight 442.7
Physical Form Powder
Chemical Name or Material Betulin
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.