Learn More
Medchemexpress LLC Betulin diacetate | 1721-69-3 | MFCD00017378 | 95.0% | 526.79 | 5 MG
SDP
Catalog No. 5000426315
Encompass_Preferred
Betulin diacetate, a triterpene and derivative of Betulin, is an anti-AID agent and also possesses anti-cancer activity.
  • Is a triterpene and derivative of Betulin
  • Acts as an anti-AID agent
  • Possesses anti-cancer activity
  • Shows cytotoxicity against various human cell lines including 518A2, A-375, A2780, A549, HT-29, HeLa, and MCF7
  • Induces melanogenesis in mouse B16 2F2 cells
  • Inhibits growth in mouse B16 2F2 cells
  • Exhibits anti-HIV activity against acutely infected H9 lymphocytes
Catalog No. 50-004-26315 Supplier Medchemexpress LLC Supplier No. HYN94375MG
Betulin diacetate, a triterpene and derivative of Betulin, is an anti-AID agent and also possesses anti-cancer activity.
  • Is a triterpene and derivative of Betulin
  • Acts as an anti-AID agent
  • Possesses anti-cancer activity
  • Shows cytotoxicity against various human cell lines including 518A2, A-375, A2780, A549, HT-29, HeLa, and MCF7
  • Induces melanogenesis in mouse B16 2F2 cells
  • Inhibits growth in mouse B16 2F2 cells
  • Exhibits anti-HIV activity against acutely infected H9 lymphocytes