Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Betulinic Acid

Catalog No. 855057100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-505-7100MG 1 kg
85-505-7500MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 85-505-7100MG Supplier Sigma Aldrich Supplier No. 855057100MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Betulinic Acid

Specifications

CAS 24634-61-5
Molecular Formula C6H7KO2
Linear Formula CH3CH=CHCH=CHCOOK
MDL Number MFCD00016546
Quantity 1 kg
Synonym 3beta-Hydroxy-20(29)-lupaene-28-oic acid; Lupatic acid; Mairin
Molecular Weight (g/mol) 150.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.