Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Betulinic Acid

Catalog No. 855057500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
85-505-7500MG 100 mg
85-505-7100MG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 85-505-7500MG Supplier Sigma Aldrich Supplier No. 855057500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Betulinic Acid

Specifications

CAS 472-15-1
Melting Point 295°C to 298°C
Molecular Formula C30H48O3
Linear Formula C30H48O3
MDL Number MFCD00009619
Quantity 100 mg
Synonym 3beta-Hydroxy-20(29)-lupaene-28-oic acid; Lupatic acid; Mairin
Molecular Weight (g/mol) 456.7 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.