Learn More
Apexbio Technology LLC Betulinic acid(Synonyms: Betulic acid, 3β-Hydroxy-lup-20(29)-en-28-oic acid, Lupatic acid, Betulonic acid), 200mg, CAS: 472-15-1.
SDP
Catalog No. 5000060318
Encompass_Preferred
Betulinic acid (CAS 472-15-1) is a pentacyclic triterpenoid obtained from natural sources exhibiting pharmacological properties relevant to oncology and infectious diseases research It induces apoptosis selectively in tumor cells by engaging mitochondrial apoptotic pathways independently of both p53 and CD95 signaling Additionally betulinic acid functions as an agonist for the bile acid receptor TGR5 demonstrating activity with an EC50 of approximately 1 04 M Due to its apoptotic effects and antiviral properties betulinic acid is extensively utilized in preclinical studies exploring cancer therapeutics and anti-HIV strategies
Catalog No. 50-000-60318 Supplier Apexbio Technology LLC Supplier No. A3237200
Betulinic acid (CAS 472-15-1) is a pentacyclic triterpenoid obtained from natural sources exhibiting pharmacological properties relevant to oncology and infectious diseases research It induces apoptosis selectively in tumor cells by engaging mitochondrial apoptotic pathways independently of both p53 and CD95 signaling Additionally betulinic acid functions as an agonist for the bile acid receptor TGR5 demonstrating activity with an EC50 of approximately 1 04 M Due to its apoptotic effects and antiviral properties betulinic acid is extensively utilized in preclinical studies exploring cancer therapeutics and anti-HIV strategies