Learn More
Cayman Chemical Betulinic Acid, Purity: ≥97%, CAS: 472-15-1, 250 mg
SDP
Catalog No. 502952196
Encompass_Preferred
An agonist of the bile acid receptor TGR5 (EC50 = 1 µM) and blocks fatty liver development in cells and mice given ethanol; reduces NF-κB signaling and expression of miR-33 in LPS-treated macrophages, increasing levels of ABCA1; inhibits replication of HIV-1 (IC50 = 1.4 µM) and has cytotoxic effects against cancer cells
Catalog No. 50-295-2196 Supplier Cayman Chemical Supplier No. 11686250 MG
An agonist of the bile acid receptor TGR5 (EC50 = 1 µM) and blocks fatty liver development in cells and mice given ethanol; reduces NF-κB signaling and expression of miR-33 in LPS-treated macrophages, increasing levels of ABCA1; inhibits replication of HIV-1 (IC50 = 1.4 µM) and has cytotoxic effects against cancer cells