Learn More
Selleck Chemical LLC Betulinic acid S3603-10mg
SDP
Catalog No. 503109248
Encompass_Preferred
Betulinic acid (ALS-357 Lupatic acid Betulic acid) a pentacyclic triterpenoid from Syzigium claviflorum is a inhibitor of HIV-1 with EC50 of 1 4 M It s reported that Betulinic acid acts as a new inhibitor of NF- B Phase 1/2
Catalog No. 50-310-9248 Supplier Selleck Chemical LLC Supplier No. S360310MG
Betulinic acid (ALS-357 Lupatic acid Betulic acid) a pentacyclic triterpenoid from Syzigium claviflorum is a inhibitor of HIV-1 with EC50 of 1 4 M It s reported that Betulinic acid acts as a new inhibitor of NF- B Phase 1/2