Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Betulinic acid | 472-15-1 | MFCD00009619 | 456.7 g/mol | C30H48O3 | 10 MG
SDP

Catalog No. 5000243170
Encompass_Preferred

Betulinic acid is a natural pentacyclic triterpenoid supplied as an analytical reference for research and chromatographic assays. It is reported to act as a eukaryotic topoisomerase I inhibitor and has documented antiviral, anti-malarial, anti-inflammatory, and antitumor activities.

  • Intended for analytical and reference use.
  • Natural pentacyclic triterpenoid with reported antiviral and antitumor activity.
  • Molecular formula C30H48O3.
  • Molecular weight 456.7 g/mol.
  • Available as 10 mM in DMSO (1 mL) or as solid reference materials for method development.
  • Suitable as a positive control and reference standard in chromatographic assays.

Catalog No. 50-002-43170 Supplier Medchemexpress LLC Supplier No. HY10529R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Betulinic acid is a natural pentacyclic triterpenoid supplied as an analytical reference for research and chromatographic assays. It is reported to act as a eukaryotic topoisomerase I inhibitor and has documented antiviral, anti-malarial, anti-inflammatory, and antitumor activities.

  • Intended for analytical and reference use.
  • Natural pentacyclic triterpenoid with reported antiviral and antitumor activity.
  • Molecular formula C30H48O3.
  • Molecular weight 456.7 g/mol.
  • Available as 10 mM in DMSO (1 mL) or as solid reference materials for method development.
  • Suitable as a positive control and reference standard in chromatographic assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.