Learn More
Medchemexpress LLC Betulinic acid | 472-15-1 | MFCD00009619 | 99.1% | 456.70 g/mol | C30H48O3 | 1 ML
SDP
Catalog No. 5000244104
Encompass_Preferred
Betulinic acid is a naturally occurring pentacyclic triterpenoid provided as a 10 mM solution in DMSO (1 mL) for laboratory research. It is commonly used in studies of apoptosis, cancer, inflammation, and autophagy due to its reported bioactivities.
  • Natural pentacyclic triterpenoid with reported anticancer and apoptotic activity.
  • Supplied as a 10 mM solution in DMSO for convenient handling and dosing.
  • High purity (99.1%) suitable for analytical and biological assays.
  • Available in multiple solid sizes for scale-up or formulation work.
  • Intended for research use only.
Catalog No. 50-002-44104 Supplier Medchemexpress LLC Supplier No. HY1052910MM 1ML
Betulinic acid is a naturally occurring pentacyclic triterpenoid provided as a 10 mM solution in DMSO (1 mL) for laboratory research. It is commonly used in studies of apoptosis, cancer, inflammation, and autophagy due to its reported bioactivities.
  • Natural pentacyclic triterpenoid with reported anticancer and apoptotic activity.
  • Supplied as a 10 mM solution in DMSO for convenient handling and dosing.
  • High purity (99.1%) suitable for analytical and biological assays.
  • Available in multiple solid sizes for scale-up or formulation work.
  • Intended for research use only.