Learn More
Medchemexpress LLC Betulinic acid | 472-15-1 | MFCD00009619 | 99.1% | 456.70 g/mol | C30H48O3 | 200 MG
SDP
Catalog No. 5000243900
Encompass_Preferred
Betulinic acid is a naturally occurring pentacyclic triterpenoid used in research as an apoptosis inducer and bioactive small molecule. It is commonly used in pharmacology and cell biology to probe apoptotic pathways, evaluate anti-inflammatory and antiviral activity, and as a scaffold for synthetic derivatization.
  • Natural pentacyclic triterpenoid compound.
  • Functions as an apoptosis inducer in cellular assays.
  • High purity suitable for research (≈99.1%).
  • Available in multiple pack sizes for screening and synthesis workflows.
  • Soluble in DMSO for preparation of stock solutions.
Catalog No. 50-002-43900 Supplier Medchemexpress LLC Supplier No. HY10529200MG
Betulinic acid is a naturally occurring pentacyclic triterpenoid used in research as an apoptosis inducer and bioactive small molecule. It is commonly used in pharmacology and cell biology to probe apoptotic pathways, evaluate anti-inflammatory and antiviral activity, and as a scaffold for synthetic derivatization.
  • Natural pentacyclic triterpenoid compound.
  • Functions as an apoptosis inducer in cellular assays.
  • High purity suitable for research (≈99.1%).
  • Available in multiple pack sizes for screening and synthesis workflows.
  • Soluble in DMSO for preparation of stock solutions.