Learn More
Medchemexpress LLC Betulinic acid | 472-15-1 | MFCD00009619 | 99.2% | 456.7 g/mol | C30H48O3 | 100 MG
SDP
Catalog No. 5000244085
Encompass_Preferred
Betulinic acid is a natural pentacyclic triterpenoid (CAS 472-15-1) provided as an analytical reference standard for research and analytical applications. It is used as a qualitative and quantitative standard in method development and validation for HPLC, GC, and MS, and is reported to have biological activities including anti-HIV, anti-malarial, anti-inflammatory, and anti-tumor effects.
  • High reported purity for analytical applications.
  • Suitable for HPLC, GC, and MS method development.
  • Intended for qualitative and quantitative reference standards.
  • Supplied as a solid reference material with defined sizes.
  • Supports research into multiple biological activities.
Catalog No. 50-002-44085 Supplier Medchemexpress LLC Supplier No. HY10529R100MG
Betulinic acid is a natural pentacyclic triterpenoid (CAS 472-15-1) provided as an analytical reference standard for research and analytical applications. It is used as a qualitative and quantitative standard in method development and validation for HPLC, GC, and MS, and is reported to have biological activities including anti-HIV, anti-malarial, anti-inflammatory, and anti-tumor effects.
  • High reported purity for analytical applications.
  • Suitable for HPLC, GC, and MS method development.
  • Intended for qualitative and quantitative reference standards.
  • Supplied as a solid reference material with defined sizes.
  • Supports research into multiple biological activities.