Learn More
Medchemexpress LLC Betulinic acid | 472-15-1 | MFCD00009619 | 99.2% | 456.70 | C30H48O3 | 25 MG
SDP
Catalog No. 5000244090
Encompass_Preferred
Betulinic acid is a natural pentacyclic triterpenoid supplied as a high-purity analytical standard for research and laboratory assays. It is reported to inhibit eukaryotic topoisomerase I (IC50 ≈ 5 μM) and has documented anti-HIV, antimalarial, anti-inflammatory, and antitumor activities. The material is intended for qualitative and quantitative analysis in HPLC, GC, and MS and is provided in small pack sizes suitable for analytical workflows.
  • High purity suitable for analytical assays.
  • Validated for HPLC, GC, and MS methodologies.
  • Provides a stable solid reference for quantitative analysis.
  • Documented biological activity including topoisomerase I inhibition.
  • Available in small pack sizes for laboratory use.
Catalog No. 50-002-44090 Supplier Medchemexpress LLC Supplier No. HY10529R25MG
Betulinic acid is a natural pentacyclic triterpenoid supplied as a high-purity analytical standard for research and laboratory assays. It is reported to inhibit eukaryotic topoisomerase I (IC50 ≈ 5 μM) and has documented anti-HIV, antimalarial, anti-inflammatory, and antitumor activities. The material is intended for qualitative and quantitative analysis in HPLC, GC, and MS and is provided in small pack sizes suitable for analytical workflows.
  • High purity suitable for analytical assays.
  • Validated for HPLC, GC, and MS methodologies.
  • Provides a stable solid reference for quantitative analysis.
  • Documented biological activity including topoisomerase I inhibition.
  • Available in small pack sizes for laboratory use.