Learn More
TARGETMOL CHEMICALS INC Betulinic acid 500MG
SDP
Catalog No. 502879952
Encompass_Preferred
Also available in 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Betulinic acid (ALS-357) is a pentacyclic lupane-type triterpene derivative of betulin (isolated from the bark of Betula alba, the common white birch) which has antiretroviral, antimalarial, and anti-inflammatory properties, as well as a more recently discovered potential as an anticancer agent, by inhibition of topoisomerase. Purity ≥98%
Catalog No. 50-287-9952 Supplier TARGETMOL CHEMICALS INC Supplier No. T2830500MG
Also available in 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Betulinic acid (ALS-357) is a pentacyclic lupane-type triterpene derivative of betulin (isolated from the bark of Betula alba, the common white birch) which has antiretroviral, antimalarial, and anti-inflammatory properties, as well as a more recently discovered potential as an anticancer agent, by inhibition of topoisomerase. Purity ≥98%