Learn More
Enzo Life Sciences Betulinic acid (~95%) (1g). CAS: 472-15-1
SDP
Catalog No. 502008834
Encompass_Preferred
Antitumor and anti-HIV agent. Induces apoptosis by activating mitochondrial permeability transition (MPT). Inhibits NF-κB activation and NF-κB-regulated gene expression induced by carcinogens and inflammatory stimuli. Decreases expression of Bcl-2 and cyclin D1. Potent proteasome activator. Alternative name: 3beta-Hydroxy-20(29)-lupaene-28-oic acid. Purity: ≥90% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO. Long Term Storage: +4°C.
Catalog No. 50-200-8834 Supplier Enzo Life Sciences Supplier No. ALX350298G001
Antitumor and anti-HIV agent. Induces apoptosis by activating mitochondrial permeability transition (MPT). Inhibits NF-κB activation and NF-κB-regulated gene expression induced by carcinogens and inflammatory stimuli. Decreases expression of Bcl-2 and cyclin D1. Potent proteasome activator. Alternative name: 3beta-Hydroxy-20(29)-lupaene-28-oic acid. Purity: ≥90% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO. Long Term Storage: +4°C.