Learn More
AdipoGen Betulinic acid (>97%) (1 g)
SDP
Catalog No. 501687370
Encompass_Preferred
Betulinic acid (>97%) (1 g), Adipogen Life Sciences. Chemical. CAS: 472-15-1. Formula: C30H48O3. MW: 456.7. Isolated from Platanus acerifolia bark. Anti-HIV compound. Anticancer compound. Apoptosis inducer. Modulator of NF-kappaB. Potent DNA topoisomerase II inhibitor. Potent proteasome activator. Antimalarial compound.
Catalog No. 50-168-7370 Supplier AdipoGen Supplier No. AGCN20417G001
Betulinic acid (>97%) (1 g), Adipogen Life Sciences. Chemical. CAS: 472-15-1. Formula: C30H48O3. MW: 456.7. Isolated from Platanus acerifolia bark. Anti-HIV compound. Anticancer compound. Apoptosis inducer. Modulator of NF-kappaB. Potent DNA topoisomerase II inhibitor. Potent proteasome activator. Antimalarial compound.