Learn More
AdipoGen Betulinic acid 97
SDP
Catalog No. 502562541
Encompass_Preferred
Chemical. CAS 472-15-1. Formula C30H48O3. MW 456.7. Isolated from Platanus acerifolia bark. Anti-HIV compound. Anticancer compound. Apoptosis inducer. Modulator of NF-kappaB. Potent DNA topoisomerase II inhibitor. Potent proteasome activator. Antimalarial compound.
Catalog No. 50-256-2541 Supplier AdipoGen Supplier No. AGCN20417M500
Chemical. CAS 472-15-1. Formula C30H48O3. MW 456.7. Isolated from Platanus acerifolia bark. Anti-HIV compound. Anticancer compound. Apoptosis inducer. Modulator of NF-kappaB. Potent DNA topoisomerase II inhibitor. Potent proteasome activator. Antimalarial compound.