Learn More
AdipoGen Betulinic acid (>99%) (100 mg)
SDP
Catalog No. 502053588
Encompass_Preferred
Chemical. CAS: 472-15-1. Formula: C30H48O3. MW: 456.7. Isolated from Platanus acerifolia bark. Anti-HIV compound. Anticancer compound. Apoptosis inducer. Modulator of NF-kappaB. Potent DNA topoisomerase II inhibitor. Potent proteasome activator. Antimalarial compound.
Catalog No. 50-205-3588 Supplier AdipoGen Supplier No. AGCN20415M100
Chemical. CAS: 472-15-1. Formula: C30H48O3. MW: 456.7. Isolated from Platanus acerifolia bark. Anti-HIV compound. Anticancer compound. Apoptosis inducer. Modulator of NF-kappaB. Potent DNA topoisomerase II inhibitor. Potent proteasome activator. Antimalarial compound.